2,4-Dichloro-6-methylpyrimidin-5-ylamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-6-methylpyrimidin-5-amine, 5-Amino-2,4-dichloro-6-methylpyrimidine

A255810-1G A255810-25G A255810-5G A255810-100G A255810-10G A255810-250MG
103812-214EA 11 USD
103812-214 103852-074 77091-534 77091-536 77091-538 77250-022 77250-024 77250-026
2,4-Dichloro-6-methylpyrimidin-5-ylamine 97%
2,4-Dichloro-6-methylpyrimidin-5-ylamine

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N₃
MW: 178.02 g/mol
Boiling Pt: 281.6 °C (760 mm Hg)
Melting Pt: 108…110 °C
Flash Pt: 124.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062586
CAS Number: 13162-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR