2,4-Dichloro-6-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A197812-1G
A197812-10G
A197812-250MG
A197812-25G
103816-968EA
7
USD
103816-968
103847-590
77085-602
77248-802
77248-804
77302-570
2,4-Dichloro-6-methylpyridine 98%
2,4-Dichloro-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅Cl₂N MW: 162.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08669834 CAS Number: 42779-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...