2,4-Dichloro-6-methoxy-quinoline 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-6-methoxyquinoline

A446998-100MG A446998-1G
103827-752EA 206 USD
103827-752 103838-818 77116-178 77307-348
2,4-Dichloro-6-methoxy-quinoline 98%
2,4-Dichloro-6-methoxy-quinoline

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂NO
MW: 228.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03285967
CAS Number: 70049-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR