2,4-Dichloro-6-methoxyquinazoline 97%
Supplier: Ambeed
|
Warning
|
A323384-1G
A323384-100MG
A323384-250MG
103825-336EA
36
USD
103825-336
103836-272
76839-190
77251-184
77251-186
2,4-Dichloro-6-methoxyquinazoline 97%
2,4-Dichloro-6-methoxyquinazoline
Precaución: for Research Use Only
Formula:
C₉H₆Cl₂N₂O MW: 229.0627 g/mol Boiling Pt: 315.9 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09954880 CAS Number: 105763-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...