2,4-Dichloro-6-methoxypyrimidine 97%
Supplier: Ambeed
|
Warning
|
A122155-100MG
A122155-10G
A122155-250MG
A122155-1G
A122155-5G
103800-180EA
21
USD
103800-180
77026-474
77026-476
77026-478
77312-314
77312-316
2,4-Dichloro-6-methoxypyrimidine 97%
2,4-Dichloro-6-methoxypyrimidine
Precaución: for Research Use Only
Formula:
C₅H₄Cl₂N₂O MW: 179.0000 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00955635 CAS Number: 43212-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...