2,4-Dichloro-6-methoxyaniline 95%

Supplier: Ambeed
Warning

A748871-1G A748871-250MG
103840-838EA 289 USD
103840-838 76845-068 76879-980
2,4-Dichloro-6-methoxyaniline 95%
2,4-Dichloro-6-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.04 g/mol
Boiling Pt: 273.9 °C (760 mm Hg)
Flash Pt: 119.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20693704
CAS Number: 93839-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR