2,4-Dichloro-6-methoxy-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A292833-1G A292833-5G A292833-10G A292833-25G A292833-100G
103813-956EA 8 USD
103813-956 103835-096 76837-698 76837-700 77250-708 77305-620 77746-546
2,4-Dichloro-6-methoxy-1,3,5-triazine 97%
2,4-Dichloro-6-methoxy-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₄H₃Cl₂N₃O
Melting Pt: 86…88 °C
Flash Pt: 157.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00128159
CAS Number: 3638-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR