2,4-Dichloro-6-fluoroquinazoline 97%

Supplier: Ambeed
Warning

A120962-250MG A120962-1G A120962-5G
103800-018EA 22 USD
103800-018 77246-108 77297-802 77297-804
2,4-Dichloro-6-fluoroquinazoline 97%
2,4-Dichloro-6-fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈H₃Cl₂FN₂
MW: 217.0300 g/mol
Boiling Pt: 201.7 °C (760 mm Hg)
Flash Pt: 75.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09954884
CAS Number: 134517-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR