2,4-Dichloro-6,7-dimethoxyquinoline 98%

Supplier: Ambeed
Warning

A567361-100MG A567361-250MG A567361-1G
103828-786EA 436 USD
103828-786 103839-928 77103-660 77103-662 77307-834
2,4-Dichloro-6,7-dimethoxyquinoline 98%
2,4-Dichloro-6,7-dimethoxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₉Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18073200
CAS Number: 72407-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR