2,4-Dichloro-6,7-dimethoxyquinazoline 97%

Supplier: Ambeed
Warning

A136450-100G A136450-10G A136450-250MG A136450-1G A136450-25G A136450-5G
103802-302EA 19 USD
103802-302 76767-538 76767-540 76767-542 77315-194 77315-196 77315-198
2,4-Dichloro-6,7-dimethoxyquinazoline 97%
2,4-Dichloro-6,7-dimethoxyquinazoline

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂N₂O₂
Boiling Pt: 340.512 °C (760 mm Hg)
Melting Pt: 172…177 °C at N/A mmHg
Flash Pt: 159.736 °C
Storage Temperature: Ambient
MDL Number: MFCD00051733
CAS Number: 27631-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR