2,4-Dichloro-6,7-dihydrothieno[3,2-d]pyrimidine 98%

Supplier: Ambeed
Warning

A463204-250MG A463204-1G A463204-5G
103827-948EA 30 USD
103827-948 103839-024 76789-494 76842-918 76842-920
2,4-Dichloro-6,7-dihydrothieno[3,2-d]pyrimidine 98%
2,4-Dichloro-6,7-dihydrothieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂S
MW: 207.0800 g/mol
Boiling Pt: 329.4 °C (760 mm Hg)
Melting Pt: 92…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109461
CAS Number: 74901-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR