2,4-Dichloro-6-(trifluoromethyl)pyrimidine 98%

Supplier: Ambeed
Warning

A133266-100G A133266-500G A133266-25G
103801-852EA 5 USD
103801-852 77246-630 77246-632 77298-616
2,4-Dichloro-6-(trifluoromethyl)pyrimidine 98%
2,4-Dichloro-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅HCl₂F₃N₂
MW: 216.97 g/mol
Boiling Pt: 206.9 °C (760 mm Hg)
Flash Pt: 78.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09910264
CAS Number: 16097-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR