2,4-Dichloro-5-nitropyridine 98%

Supplier: Ambeed
Warning

A177678-1G A177678-25G A177678-5G A177678-100G A177678-100MG A177678-10G A177678-250MG A177678-500G
103807-258EA 26 USD
103807-258 103845-590 77082-980 77082-982 77082-984 77163-756 77163-758 77163-760 77163-762 77892-170
2,4-Dichloro-5-nitropyridine 98%
2,4-Dichloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
Boiling Pt: 282.329 °C (760 mm Hg)
Melting Pt: 40…44 °C
Flash Pt: 124.548 °C
Storage Temperature: Ambient
MDL Number: MFCD07368834
CAS Number: 4487-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR