2,4-Dichloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A134128-100MG A134128-250MG A134128-1G A134128-5G
103801-978EA 36 USD
103801-978 76782-526 76782-528 77298-692 77298-694
2,4-Dichloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 97%
2,4-Dichloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N₃
MW: 202.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755925
CAS Number: 129872-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR