2,4-Dichloro-5-methoxyaniline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2,4-dichloroanisole, 4,6-Dichloro-m-anisidine
A226967-25G
A226967-5G
A226967-100G
A226967-10G
A226967-1G
103819-284EA
7
USD
103819-284
103850-192
77088-942
77088-944
77152-820
77152-822
77303-786
2,4-Dichloro-5-methoxyaniline 98%
2,4-Dichloro-5-methoxyaniline
Precaución: for Research Use Only
Formula:
C₇H₇Cl₂NO Boiling Pt: 290.1 °C (760 mm Hg) Melting Pt: 51 °C Flash Pt: 129.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00974410 CAS Number: 98446-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...