2,4-Dichloro-5-iodopyrimidine 97%

Supplier: Ambeed
Danger

A258764-5G A258764-1G A258764-10G
103812-396EA 9 USD
103812-396 103852-274 77091-808 77304-748 77683-789
2,4-Dichloro-5-iodopyrimidine 97%
2,4-Dichloro-5-iodopyrimidine

Precaución: For research use only.

Formula: C₄HCl₂IN₂
MW: 274.87 g/mol
Melting Pt: 65…69 °C
Flash Pt: 141.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01898087
CAS Number: 13544-44-0
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR