2,4-Dichloro-5-iodopyridine 98%

Supplier: Ambeed
Warning

A131812-100MG A131812-10G A131812-250MG A131812-1G A131812-5G
103801-638EA 18 USD
103801-638 76782-392 76782-394 76782-396 77314-168 77314-170
2,4-Dichloro-5-iodopyridine 98%
2,4-Dichloro-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂IN
Boiling Pt: 282.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09033755
CAS Number: 343781-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR