2,4-Dichloro-5-ethylpyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-5-ethyl-1,3-diazine

A213378-250MG A213378-1G A213378-5G
103818-296EA 53 USD
103818-296 103849-004 76975-594 77087-518 77303-284
2,4-Dichloro-5-ethylpyrimidine 97%
2,4-Dichloro-5-ethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.031 g/mol
Boiling Pt: 246 °C (760 mm Hg)
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11040247
CAS Number: 34171-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR