2,4-Dichloro-5-ethoxypyrimidine 95%

Supplier: Ambeed
Warning

A164719-100MG A164719-250MG A164719-1G
103805-892EA 138 USD
103805-892 103844-150 76985-200 76985-202 77300-730
2,4-Dichloro-5-ethoxypyrimidine 95%
2,4-Dichloro-5-ethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂O
MW: 193.03 g/mol
Boiling Pt: 273.7 °C (760 mm Hg)
Melting Pt: 41…42 °C
Flash Pt: 119.4 °C
Storage Temperature: Ambient
MDL Number: MFCD21648476
CAS Number: 280582-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR