2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine 97%

Supplier: Ambeed
Warning

A168340-100MG A168340-250MG A168340-1G A168340-5G
103806-280EA 24 USD
103806-280 103844-538 77028-914 77028-916 77300-928 77300-930
2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine 97%
2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MW: 191.0200 g/mol
Boiling Pt: 337.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD13250060
CAS Number: 848398-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR