2,4-Dichloro-3-nitropyridine 98%

Supplier: Ambeed
Warning

A133365-100G A133365-10G A133365-500G A133365-1G A133365-25G A133365-5G
103801-864EA 5 USD
103801-864 77246-634 77246-636 77246-638 77298-624 77314-508 77314-510
2,4-Dichloro-3-nitropyridine 98%
2,4-Dichloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MW: 192.9876 g/mol
Boiling Pt: 285.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09261088
CAS Number: 5975-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR