2,4-Dichloro-3-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
A109852-10G
A109852-250MG
A109852-1G
A109852-5G
103797-578EA
250
USD
103797-578
103823-382
76995-144
76995-146
77283-900
77310-186
2,4-Dichloro-3-methylpyridine 97%
2,4-Dichloro-3-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅Cl₂N MW: 162.0200 g/mol Boiling Pt: 218.7±35.0 °C (760 mm Hg) Melting Pt: 40…43 °C Flash Pt: 106.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09263623 CAS Number: 132097-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...