2,4-Dichloro-3-fluoropyridine 97%
Supplier: Ambeed
|
Warning
|
A137487-100MG
A137487-250MG
A137487-5G
103802-460EA
54
USD
103802-460
76811-598
76811-600
77315-426
2,4-Dichloro-3-fluoropyridine 97%
2,4-Dichloro-3-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₂Cl₂FN Boiling Pt: 187.7±35.0 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD12196994 CAS Number: 628691-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...