1,3-Dichloro-2-fluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A315855-1G A315855-25G A315855-5G
103825-088EA 12 USD
103825-088 103836-010 76838-840 76838-842 76838-844
1,3-Dichloro-2-fluoro-4-nitrobenzene 98%
1,3-Dichloro-2-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FNO₂
Boiling Pt: 279.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00042240
CAS Number: 393-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR