2,4-Dichloro-3-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

A217765-1G A217765-100MG A217765-250MG
103818-650EA 120 USD
103818-650 77088-020 77192-826 77192-828
2,4-Dichloro-3-(trifluoromethyl)aniline 97%
2,4-Dichloro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD28787614
CAS Number: 1804908-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR