2,4-Dichloro-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A762574-1G A762574-5G A762574-250MG
103840-894EA 22 USD
103840-894 76845-122 76845-124 77105-346
2,4-Dichloro-1,3,5-triazine 97%
2,4-Dichloro-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₃HCl₂N₃
Boiling Pt: 329.4 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD04115347
CAS Number: 2831-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR