2,4-Dibromothiazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A153924-100MG A153924-250MG A153924-1G A153924-5G A153924-10G
103804-582EA 35 USD
103804-582 103842-762 76783-512 76783-514 77318-450 77318-452 77871-763
2,4-Dibromothiazole-5-carbaldehyde 97%
2,4-Dibromothiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₄HBr₂NOS
Boiling Pt: 317 °C (760 mm Hg)
Flash Pt: 145.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04117944
CAS Number: 139669-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR