2,4-Dibromothiazole 98%

Supplier: Ambeed
Warning

A135340-100G A135340-10G A135340-500G A135340-5G A135340-1G A135340-25G
103802-136EA 5 USD
103802-136 77027-272 77027-274 77027-276 77298-770 77314-948 77314-950
2,4-Dibromothiazole 98%
2,4-Dibromothiazole

Precaución: for Research Use Only

Formula: C₃HBr₂NS
Boiling Pt: 242.8 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 100.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01318994
CAS Number: 4175-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR