1-Amino-2,4-dibromonaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dibromonaphthalen-1-amine, 2,4-Dibromo-1-naphthylamine

A161449-10G A161449-250MG A161449-1G A161449-25G A161449-5G
103805-492EA 16 USD
103805-492 103843-730 77150-906 77150-908 77319-712 77319-714 77319-716
1-Amino-2,4-dibromonaphthalene 97%
1-Amino-2,4-dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇Br₂N
MW: 300.977 g/mol
Boiling Pt: 369.7 °C (760 mm Hg)
Melting Pt: 120…124 °C
Flash Pt: 177.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00155345
CAS Number: 20191-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR