2,4-Dibromobenzonitrile 98%

Supplier: Ambeed
Warning

A244882-250MG A244882-1G A244882-5G
103811-606EA 43 USD
103811-606 103851-440 77140-358 77304-334 77304-336
2,4-Dibromobenzonitrile 98%
2,4-Dibromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Br₂N
MW: 260.91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185491
CAS Number: 78222-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR