2,4-Dibromo-5-nitropyridine 98%

Supplier: Ambeed
Warning

A434999-10G A434999-250MG
103827-584EA 18 USD
103827-584 103838-650 77746-884 77746-885
2,4-Dibromo-5-nitropyridine 98%
2,4-Dibromo-5-nitropyridine

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₅H₂Br₂N₂O₂
MW: 281.8900 g/mol
Boiling Pt: 292 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09832226
CAS Number: 4487-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR