2,4-Dibromo-5-methylpyridine 98%

Supplier: Ambeed
Warning

A101186-250MG A101186-25G A101186-10G
103795-846EA 57 USD
103795-846 103821-530 76869-134 76869-136 77570-092
2,4-Dibromo-5-methylpyridine 98%
2,4-Dibromo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.91 g/mol
Boiling Pt: 276.4 °C (760 mm Hg)
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100634
CAS Number: 79055-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR