1,3-Dibromo-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A156388-100G A156388-10G A156388-250MG A156388-500G A156388-1G A156388-25G A156388-5G
103804-870EA 6 USD
103804-870 103843-070 77247-484 77247-486 77247-488 77247-490 77318-838 77318-840 77318-842
1,3-Dibromo-4-nitrobenzene 97%
1,3-Dibromo-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
Boiling Pt: 270 °C (760 mm Hg)
Melting Pt: 61…62 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD02660416
CAS Number: 51686-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR