2,4-Bis(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A287523-5G A287523-100G A287523-10G A287523-25G
103813-762EA 10 USD
103813-762 103834-904 76837-458 77014-708 77014-710 77305-516
2,4-Bis(trifluoromethyl)phenylboronic acid 98%
2,4-Bis(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₅BF₆O₂
Boiling Pt: 245.4 °C (760 mm Hg)
Melting Pt: 110…117 °C
Flash Pt: 102.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01631349
CAS Number: 153254-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR