2,4'-Bipyridyl 97%
Supplier: Ambeed
|
Warning
|
A215432-25G
A215432-1G
A215432-100G
103818-486EA
10
USD
103818-486
103849-192
76800-748
77087-720
77879-323
2,4'-Bipyridyl 97%
2,4'-Bipyridyl
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂ Boiling Pt: 280.8 °C (760 mm Hg) Melting Pt: 58…62 °C (760 mm Hg) Flash Pt: 105.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006217 CAS Number: 581-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...