2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate] 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis(3-(3-(tert-butyl)-4-hydroxy-5-methylphenyl)propanoate)

A141707-100G A141707-10G A141707-500G A141707-1G A141707-25G A141707-5G
103803-044EA 8 USD
103803-044 76782-960 76782-962 76782-964 77316-212 77316-214 77316-216
2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate] 98%
2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diylbis(2-methylpropane-2,1-diyl) bis[3-[3-(tert-butyl)-4-hydroxy-5-methylphenyl]propanoate]

Precaución: for Research Use Only

Formula: C₄₃H₆₄O₁₀
MW: 740.98 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09838745
CAS Number: 90498-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR