2,4,7-Trichloropyrido[2,3-d]pyrimidine 95%

Supplier: Ambeed
Warning

A245594-100MG A245594-250MG A245594-1G
103811-656EA 114 USD
103811-656 103851-488 76786-668 76786-670 77304-362
2,4,7-Trichloropyrido[2,3-d]pyrimidine 95%
2,4,7-Trichloropyrido[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₂Cl₃N₃
Boiling Pt: 316.4±42.0 °C (760 mm Hg)
Flash Pt: 174.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109463
CAS Number: 938443-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR