2,4,6-Tri-tert-butylpyrimidine 98%

Supplier: Ambeed
Warning

A599304-1G A599304-25G A599304-5G A599304-100G A599304-10G
103828-970EA 5 USD
103828-970 103840-106 76844-258 76844-260 76844-262 77242-564 77242-566
2,4,6-Tri-tert-butylpyrimidine 98%
2,4,6-Tri-tert-butylpyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD03094045
CAS Number: 67490-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR