2,4,6-Tris(4-bromophenyl)-1,3,5-triazine 95%

Supplier: Ambeed
Warning

A103698-100G A103698-250MG A103698-25G A103698-1G A103698-5G
103796-598EA 12 USD
103796-598 103822-306 77093-612 77093-614 77093-616 77309-174 77309-176
2,4,6-Tris(4-bromophenyl)-1,3,5-triazine 95%
2,4,6-Tris(4-bromophenyl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₂Br₃N₃
MW: 546.06 g/mol
Boiling Pt: 633.4 °C (760 mm Hg)
Flash Pt: 336.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04116312
CAS Number: 30363-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR