2,4,6-Trimethylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane

A107951-100G A107951-10G A107951-25G A107951-5G A107951-1G
103797-274EA 5 USD
103797-274 103823-048 77245-528 77245-530 77245-532 77283-670 77309-888
2,4,6-Trimethylphenylboronic acid 97%
2,4,6-Trimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₂
MW: 164.01
Boiling Pt: 309.1 °C (760 mm Hg)
Melting Pt: 115…122 °C
Flash Pt: 140.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00236060
CAS Number: 5980-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR