2-Iodomesitylene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4,6-Trimethyliodobenzene, 1-Iodo-2,4,6-trimethylbenzene, 2-Iodo-1,3,5-trimethylbenzene, Iodomesitylene
A175743-5G
A175743-100G
A175743-10G
A175743-25G
103845-396EA
9
USD
103845-396
77082-740
77248-134
77248-136
77248-138
2-Iodomesitylene 98%
2-Iodomesitylene
Precaución: for Research Use Only
Formula:
C₉H₁₁I MW: 246.09 g/mol Boiling Pt: 244.9 °C (760 mm Hg) Melting Pt: 28…32 °C Flash Pt: 107.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013707 CAS Number: 4028-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...