2-Bromomesitylene 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene

A100341-100G A100341-10G A100341-25G A100341-500G A100341-1000G
103807-756EA 161 USD
103807-756 77107-148 77107-150 77107-152 77107-154 77296-644
2-Bromomesitylene 98%
2-Bromomesitylene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 225 °C (760 mm Hg)
Melting Pt: 2 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000073
CAS Number: 576-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR