2,4,6-Trimethylbenzonitrile 98%

Supplier: Ambeed
Danger

A201654-1G A201654-5G A201654-10G
103817-312EA 16 USD
103817-312 103847-954 77086-030 77086-032 77248-856
2,4,6-Trimethylbenzonitrile 98%
2,4,6-Trimethylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₁N
MW: 145.2 g/mol
Boiling Pt: 260.3±9.0 °C (760 mm Hg)
Melting Pt: 55 °C
Flash Pt: 111.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00016378
CAS Number: 2571-52-0
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR