2,4,6-Trimethylbenzaldehyde 95%
Supplier: Ambeed
|
Warning
|
A196878-100G
A196878-10G
A196878-500G
A196878-25G
A196878-5G
103847-486EA
5
USD
103847-486
77112-002
77112-004
77112-006
77302-532
77302-534
2,4,6-Trimethylbenzaldehyde 95%
2,4,6-Trimethylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O Boiling Pt: 241.5 °C (760 mm Hg) Melting Pt: 10…12 °C Flash Pt: 105.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003341 CAS Number: 487-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...