2,4,6-Trimethyl-m-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Diamino-2,4,6-trimethylbenzene, 2,4-Diamino-1,3,5-trimethylbenzene, 2,4,6-Trimethyl-1,3-phenylenediamine, 2,4-Diaminomesitylene, 2,4,6-Trimethylbenzene-1,3-diamine

A179844-25G A179844-5G A179844-100G
103845-832EA 7 USD
103845-832 77301-650 77301-652 77746-172
2,4,6-Trimethyl-m-phenylenediamine 98%
2,4,6-Trimethyl-m-phenylenediamine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
Boiling Pt: 297.7 °C (760 mm Hg)
Melting Pt: 89…91 °C
Flash Pt: 158.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00010149
CAS Number: 3102-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR