2,4,6-Trimethoxyaniline 98%

Supplier: Ambeed
Warning

A478731-1G A478731-100MG A478731-250MG A478731-5G
103828-104EA 68 USD
103828-104 103839-176 76843-118 77253-122 77253-124 77307-528
2,4,6-Trimethoxyaniline 98%
2,4,6-Trimethoxyaniline

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Boiling Pt: 306.9 °C (760 mm Hg)
Flash Pt: 152.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00017165
CAS Number: 14227-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR