2,4,6-Triformylphloroglucinol 95%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Trihydroxybenzene-1,3,5-tricarbaldehyde

A227726-5G A227726-100MG A227726-250MG A227726-25G
103819-338EA 27 USD
103819-338 103850-246 77089-030 77237-770 77568-212 77746-335
2,4,6-Triformylphloroglucinol 95%
2,4,6-Triformylphloroglucinol

Precaución: For research use only.

Formula: C₉H₆O₆
MW: 210.14 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Melting Pt: 198…200 °C
Flash Pt: 109.5 °C
Storage Temperature: Freezer
MDL Number: MFCD24713435
CAS Number: 34374-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR