2,4,6-Trifluoropyridine 95%

Supplier: Ambeed
Danger

A734190-1G A734190-5G A734190-10G
103840-800EA 98 USD
103840-800 76845-026 76845-028 77105-124
2,4,6-Trifluoropyridine 95%
2,4,6-Trifluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂F₃N
Boiling Pt: 121.9 °C (760 mm Hg)
Flash Pt: 17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001161
CAS Number: 3512-17-2
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR