2,4,6-Trifluorobenzoic acid 95%

Supplier: Ambeed
Warning

A907777-25G A907777-5G A907777-100G A907777-10G A907777-1G
103841-364EA 12 USD
103841-364 76845-638 76845-640 77118-812 77118-814 77118-816
2,4,6-Trifluorobenzoic acid 95%
2,4,6-Trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Boiling Pt: 218.2±35.0 °C (760 mm Hg)
Melting Pt: 142…145 °C
Flash Pt: 85.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00042398
CAS Number: 28314-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR