2,4,6-Trichloropyrimidine-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Trichloro-5-formylpyrimidine, 2,4,6-Trichloro-5-pyrimidinecarboxaldehyde

A104459-10G A104459-25G A104459-1G A104459-5G A104459-100G
103796-712EA 10 USD
103796-712 103822-426 77093-638 77093-640 77283-210 77309-292 77548-992
2,4,6-Trichloropyrimidine-5-carbaldehyde 97%
2,4,6-Trichloropyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅HCl₃N₂O
MW: 211.43 g/mol
Boiling Pt: 278.6 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD02257700
CAS Number: 50270-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR